missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Dideoxyinosine, For HPLC analysis, 101.0%, MP Biomedicals™

Product Code. 11482292 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482292 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11482292 Supplier MP Biomedicals Supplier No. 0215088125
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 69655-05-6
Color White
Assay Percent Range 101%
Molecular Formula C10H12N4O3
MDL Number MFCD00077728
Quantity 25 mg
Synonym didanosine, 2',3'-dideoxyinosine, dideoxyinosine, videx, videx ec, ddino, inosine, 2',3'-dideoxy, didanosina, didanosinum, ddi
InChI Key BXZVVICBKDXVGW-JHPDDGAFNA-N
SMILES OC[C@@H]1CC[C@@H](O1)N1C=NC2=C1N=CNC2=O
IUPAC Name 9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Molecular Weight (g/mol) 236.23
PubChem CID 50599
ChEBI CHEBI:490877
Formula Weight 236.23
Physical Form Powder
Chemical Name or Material 2', 3'-Dideoxyinosine, For HPLC analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.