Learn More
2,3-Dichloropropionyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2,3-Dichloropropionyl Chloride |
| CAS | 7623-13-4 |
| Molecular Formula | C3H3Cl3O |
| Quantity | 1 g |
| Synonym | 2,3-Dichloropropanoyl Chloride, α,β-Dichloropropionic Acid Chloride, α,β-Dichloropropionyl Chloride |
| InChI Formula | InChI=1S/C3H3Cl3O/c4-1-2(5)3(6)7/h2H,1H2 |
| SMILES | C(C(=O)Cl)(CCl)Cl |
| IUPAC Name | 2,3-dichloropropanoyl chloride |
| Molecular Weight (g/mol) | 161.41 |
| Formula Weight | 159.9249 |