missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromophenol, TRC

Product Code. 30135081
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135081 1 g 1g
30010255 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135081 Supplier Toronto Research Chemicals Supplier No. TRCD4257401G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57383-80-9
Molecular Formula C6H4Br2O
Molecular Weight (g/mol) 251.9
Synonym Phenol, 2,3-dibromo-, 2,3-Dibromophenol
SMILES Oc1cccc(Br)c1Br

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H4Br2O/c7-4-2-1-3-5(9)6(4)8/h1-3,9H
Formula Weight 249.8629
Chemical Name or Material 2,3-Dibromophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.