Learn More
2,3-Dibromo-3-(2-bromophenyl)propionic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2,3-Dibromo-3-(2-bromophenyl)propionic Acid |
| CAS | 246876-04-0 |
| Molecular Formula | C9 H7 Br3 O2 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C9H7Br3O2/c10-6-4-2-1-3-5(6)7(11)8(12)9(13)14/h1-4,7-8H,(H,13,14) |
| SMILES | OC(=O)C(Br)C(Br)c1ccccc1Br |
| IUPAC Name | 2,3-dibromo-3-(2-bromophenyl)propanoic acid |
| Molecular Weight (g/mol) | 386.86 |
| Formula Weight | 383.7996 |