missing translation for 'onlineSavingsMsg'
Learn More

2,3-Bis(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Product Code. 11368746
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368746 1 g 1g
11378746 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368746 Supplier Thermo Scientific Alfa Aesar Supplier No. L19445.03
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1644-68-4
Molecular Formula C7H3F6N
Molecular Weight (g/mol) 215.098
MDL Number MFCD03093763
InChI Key RRNXYHYDSDAOFW-UHFFFAOYSA-N
Synonym 2,3-bis trifluoromethyl pyridine, pyridine, 2,3-bis trifluoromethyl, pubchem7694, acmc-209dra, bis trifluoromethyl pyridine, 2,3-bis trifluoromethyl pyridine #, pyridine,2,3-bis trifluoromethyl
PubChem CID 589865
IUPAC Name 2,3-bis(trifluoromethyl)pyridine
SMILES C1=CC(=C(N=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point -2°C
Boiling Point 85°C (18 mmHg)
Flash Point 70°C (158°F)
Quantity 1 g
UN Number UN2810
Beilstein 1622028
Formula Weight 215.1
Percent Purity 97%
Chemical Name or Material 2,3-Bis(trifluoromethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.