missing translation for 'onlineSavingsMsg'
Learn More

2,3,6-Trimethylphenol, MP Biomedicals

Product Code. 11439464 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439464 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11439464 Supplier MP Biomedicals Supplier No. 0522280310

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 2416-94-6
Molecular Formula C9H12O
MDL Number MFCD00002229
Quantity 10 g
Synonym 3-hydroxypseudocumene, phenol, 2,3,6-trimethyl, 1-hydroxy-2,3,6-trimethylbenzene, 2,3,6-trimethyl-phenol, unii-05wkl2l5lj, 2,3,6-trimethyl phenol, 05wkl2l5lj, dsstox_cid_2187, dsstox_rid_76518, dsstox_gsid_22187
InChI Key QQOMQLYQAXGHSU-UHFFFAOYSA-N
SMILES CC1=CC=C(C)C(O)=C1C
IUPAC Name 2,3,6-trimethylphenol
Molecular Weight (g/mol) 136.19
PubChem CID 17016
Formula Weight 136.19
Chemical Name or Material 2, 3, 6-Trimethyl Phenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.