missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Triphenyl Tetrazolium Chloride, MP Biomedicals

Product Code. 11443521 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443521 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11443521 Supplier MP Biomedicals Supplier No. 0210312610
  • ONLINE EXCLUSIVE PRICE
    78.30 EUR
    74.55 EUR / 10g
    Save 3.75 EUR
    4% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • 2,3,5-Triphenyltetrazolium Chloride is commonly used in biochemical experiments especially to indicate cellular respiration
  • TTC has been employed in autopsy pathology to assist identification of post-mortem myocardial infarctions
  • Healthy viable heart muscle will stain deep red from the cardiac lactate dehydrogenase while areas of potential infarctions will be more pale
  • Useful indicator for reducing substances

Specifications

CAS 298-96-4
Melting Point 243°C to 250°C (literature)
Color White
Molecular Formula C19H15ClN4
Linear Formula C19H15N4Cl
MDL Number MFCD00011963
Quantity 10 g
Synonym 2,3,5-triphenyltetrazolium chloride, tetrazolium red, uroscreen, red tetrazolium, urocheck, vitastain, tetrazolium chloride, triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride, tetrazolium chloride
Solubility Information Soluble in ethanol, alcohol, water (10mg/mL - clear, colorless solution) or acetone. Insoluble in ether.
InChI Key PKDBCJSWQUOKDO-UHFFFAOYSA-M
SMILES [Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name triphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Weight (g/mol) 334.81
PubChem CID 9283
ChEBI CHEBI:78019
Formula Weight 334.8
Physical Form Powder
Chemical Name or Material 2,3,5-TRIPHENYL TETRAZOLIUM CHLORIDE
Show More Show Less
compliance-icons
Product Identifier
  • 2,3,5-Triphenyltetrazolium Chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2,3,5-Triphenyltetrazolium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.