Learn More
2’,3’,5’-Triacetyluridine, TRC
Product Code. 30455114
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455114 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455114 Supplier Toronto Research Chemicals Supplier No. TRCT7670255G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2',3',5'-Triacetyluridine
CAS 4105-38-8
Molecular Formula C15 H18 N2 O9
Quantity 5 g
Synonym 2',3',5'-Tri-O-acetyluridine, PN 401, RG 2133, Tri-O-acetyl uridine, Uridine triacetate, Vistonuridine
InChI Formula InChI=1S/C15H18N2O9/c1-7(18)23-6-10-12(24-8(2)19)13(25-9(3)20)14(26-10)17-5-4-11(21)16-15(17)22/h4-5,10,12-14H,6H2,1-3H3,(H,16,21,22)/t10-,12-,13-,14-/m1/s1
SMILES CC(=O)OC[C@H]1O[C@H]([C@H](OC(=O)C)[C@@H]1OC(=O)C)N2C=CC(=O)NC2=O
IUPAC Name [(2R,3R,4R,5R)-3,4-diacetyloxy-5-(2,4-dioxopyrimidin-1-yl)oxolan-2-yl]methyl acetate
Molecular Weight (g/mol) 370.311
Formula Weight 370.101