missing translation for 'onlineSavingsMsg'
Learn More

2’,3’,5’-Tri-O-acetyl Guanosine-13D2,15N, TRC

Product Code. 30457202
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2.5 mg
Unit Size:
0.50mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457202 2.5 mg 2.5mg
30450403 1 mg 1mg
30450317 0.5 mg 0.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457202 Supplier Toronto Research Chemicals Supplier No. TRCT7652372.5MG
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2’,3’,5’-Tri-O-acetyl Guanosine-13C2,15N
Molecular Formula 13C2 C14 H19 15N N4 O8
Quantity 2.5 mg
Synonym 2’,3’,5’-Tri-O-acetylguanosine-13C2,15N, Guanosine Triacetate-13C2,15N, NSC 66387-13C2,15N, Triacetylguanosine-13C2,15N
InChI Formula InChI=1S/C16H19N5O8/c1-6(22)26-4-9-11(27-7(2)23)12(28-8(3)24)15(29-9)21-5-18-10-13(21)19-16(17)20-14(10)25/h5,9,11-12,15H,4H2,1-3H3,(H3,17,19,20,25)/t9-,11-,12-,15-/m1/s1/i10+1,13+1,18+1
SMILES CC(=O)OC[C@H]1O[C@H]([C@H](OC(=O)C)[C@@H]1OC(=O)C)n2c[15n][13c]3[13c]2N=C(N)NC3=O
IUPAC Name [(2R,3R,4R,5R)-3,4-diacetyloxy-5-(2-amino-6-oxo-1H-purin-9-yl)oxolan-2-yl]methyl acetate
Molecular Weight (g/mol) 412.329
Formula Weight 412.127
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.