missing translation for 'onlineSavingsMsg'
Learn More

2-(3,5-Dimethyl-1H-pyrazol-4-yl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10772273
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10772273 1 g Amber Glass Bottle 1g
10602633 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10772273 Supplier Thermo Scientific Maybridge Supplier No. CC15701DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 321309-43-7
Molecular Formula C12H12N2O2
Molecular Weight (g/mol) 216.24
MDL Number MFCD02682016
InChI Key JNHKHFADRGJMJS-UHFFFAOYSA-N
Synonym 2-3,5-dimethyl-1h-pyrazol-4-yl benzoic acid, 4-2-carboxyphenyl-3,5-dimethylpyrazole, benzoic acid,2-3,5-dimethyl-1h-pyrazol-4-yl, 2-3,5-dimethylpyrazol-4-yl benzoic acid, 2-3,5-dimethyl-1h-pyrazol-4-yl-benzoic acid
PubChem CID 2776446
IUPAC Name 2-(3,5-dimethyl-1H-pyrazol-4-yl)benzoic acid
SMILES CC1=C(C(C)=NN1)C1=CC=CC=C1C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 216.24
Percent Purity 97%
Chemical Name or Material 2-(3, 5-Dimethyl-1H-pyrazol-4-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.