Learn More
2-(3,5-Di-tert-butyl-2-hydroxyphenyl)-2H-benzotriazole, Dr. Ehrenstorfer
Product Code. 30327976
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327976 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30327976 Supplier Dr. Ehrenstorfer Supplier No. DREC12253300
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(3,5-Di-tert-butyl-2-hydroxyphenyl)-2H-benzotriazole
CAS 3846-71-7
Analyte or Component Names 2-(3,5-Di-tert-butyl-2-hydroxyphenyl)-2H-benzotriazole
Shipping Condition Room Temperature
Molecular Formula C20 H25 N3 O
Certifications/Compliance ISO 17034
Synonym Phenol, 2-(2H-benzotriazol-2-yl)-4,6-di-tert-butyl- (7CI,8CI), 2-(2H-Benzotriazol-2-yl)-4,6-bis(1,1-dimethylethyl)phenol, 2-(2-Benzotriazolyl)-4,6-di-tert-butylphenol, 2-(2-Hydroxy-3,5-di-tert-butylphenyl)-2H-benzotriazole, 2-(2-Hydroxy-3,5-di-tert-butylphenyl)benzotriazole, 2-(2'-Hydroxy-3',5'-di-t-butylphenyl)benzotriazole, 2-(2'-Hydroxy-3',5'-di-tert-butylphenyl)benzotriazole, 2-(2'-Hydroxy-3'5-di-tert-butylphenyl) benzotriazole, 2-(3,5-Di-tert-butyl-2-hydroxyphenyl)-2H-benzotriazole, 2-(3,5-Di-tert-butyl-2-hydroxyphenyl)benzotriazole, 2-(3',5'-Di-tert-butyl-2'-hydroxyphenyl)benzotriazole, Eversorb 77, Kemisorb 75, Seesorb 705, Sumisorb 320, Tinuvin 320, UV 320, Viosorb 582
InChI Formula InChI=1S/C20H25N3O/c1-19(2,3)13-11-14(20(4,5)6)18(24)17(12-13)23-21-15-9-7-8-10-16(15)22-23/h7-12,24H,1-6H3
Solution Type Neat
SMILES CC(C)(C)c1cc(c(O)c(c1)C(C)(C)C)n2nc3ccccc3n2
IUPAC Name 2-(benzotriazol-2-yl)-4,6-ditert-butylphenol
Industry Type Environmental Testing
Molecular Weight (g/mol) 323.43
Quantity 100 mg
Formula Weight 323.1998 g/mol