missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrachlorophenol, TRC

Product Code. 30135267
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135267 10 mg 10mg
30007546 25 mg 25mg
30011679 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135267 Supplier Toronto Research Chemicals Supplier No. TRCT29228510MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 935-95-5
Molecular Formula C6 H2 Cl4 O
Molecular Weight (g/mol) 231.89
Synonym Phenol, 2,3,5,6-tetrachloro-, 2,3,5,6-Tetrachlorophenol, NSC 407823
IUPAC Name 2,3,5,6-tetrachlorophenol
SMILES Oc1c(Cl)c(Cl)cc(Cl)c1Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C6H2Cl4O/c7-2-1-3(8)5(10)6(11)4(2)9/h1,11H
Formula Weight 229.886
Percent Purity >95%
Chemical Name or Material 2,3,5,6-Tetrachlorophenol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.