Learn More
2,3,4-Tribromophenol, TRC
Specifications
Specifications
| Chemical Name or Material | 2,3,4-Tribromophenol |
| CAS | 138507-65-0 |
| Molecular Formula | C6H3Br3O |
| Quantity | 10 mg |
| Synonym | Phenol','2,3,4-tribromo-, 2,3,4-Tribromophenol |
| InChI Formula | InChI=1S/C6H3Br3O/c7-3-1-2-4(10)6(9)5(3)8/h1-2,10H |
| SMILES | Oc1ccc(Br)c(Br)c1Br |
| IUPAC Name | 2,3,4-tribromophenol |
| Molecular Weight (g/mol) | 330.8 |
| Formula Weight | 327.7734 |