Learn More
2-(3,4-Dimethoxyphenyl)ethanol, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 7417-21-2 |
| Molecular Formula | C10H14O3 |
| MDL Number | MFCD00002894 |
| Quantity | 10 g |
| SMILES | COC1=CC=C(CCO)C=C1OC |
| Molecular Weight (g/mol) | 182.22 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-(3,4-Dimethoxyphenyl)ethanol |