Learn More
2,3,4,6-Tetrakis(2,2-dimethylpropanoate) D-Glucono-1,5-lactone, TRC
Specifications
Specifications
| Chemical Name or Material | 2,3,4,6-Tetrakis(2,2-dimethylpropanoate) D-Glucono-1,5-lactone |
| CAS | 157737-38-7 |
| Molecular Formula | C26H42O10 |
| Quantity | 50 mg |
| Synonym | (3R,4S,5R,6R)-2-Oxo-6-((pivaloyloxy)methyl)tetrahydro-2H-pyran-3,4,5-triyl tris(2,2-dimethylpropanoate) |
| InChI Formula | InChI=1S/C26H42O10/c1-23(2,3)19(28)32-13-14-15(34-20(29)24(4,5)6)16(35-21(30)25(7,8)9)17(18(27)33-14)36-22(31)26(10,11)12/h14-17H,13H2,1-12H3/t14-,15-,16+,17-/m1/s1 |
| SMILES | CC(C)(C)C(=O)OC[C@H]1OC(=O)[C@H](OC(=O)C(C)(C)C)[C@@H](OC(=O)C(C)(C)C)[C@@H]1OC(=O)C(C)(C)C |
| IUPAC Name | [(2R,3R,4S,5R)-3,4,5-tris(2,2-dimethylpropanoyloxy)-6-oxooxan-2-yl]methyl 2,2-dimethylpropanoate |
| Molecular Weight (g/mol) | 514.61 |
| Formula Weight | 514.2778 |