missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrafluorobenzoic acid, TRC

Product Code. 30004579
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004579 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30004579 Supplier Toronto Research Chemicals Supplier No. TRCT2929855G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1201-31-6
Molecular Formula C7 H2 F4 O2
Quantity 5 g
Synonym 2,3,4,5-Tetrafluorobenzoic Acid, 3,4,5,6-Tetrafluorobenzoic Acid, NSC 168728
InChI Formula InChI=1S/C7H2F4O2/c8-3-1-2(7(12)13)4(9)6(11)5(3)10/h1H,(H,12,13)
SMILES OC(=O)c1cc(F)c(F)c(F)c1F
IUPAC Name 2,3,4,5-tetrafluorobenzoic acid
Molecular Weight (g/mol) 194.08
Formula Weight 193.9991
Chemical Name or Material 2,3,4,5-Tetrafluorobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.