Learn More
2,3,4,5-Tetrachlorophenol, TRC
Product Code. 30138616
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138616 1 g 1g
30003996 500 mg 500mg
30005785 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138616 Supplier Toronto Research Chemicals Supplier No. TRCT2923501G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4901-51-3
Molecular Formula C6 H2 Cl4 O
Molecular Weight (g/mol) 231.89
Synonym Phenol, 2,3,4,5-tetrachloro-, 2,3,4,5-Tetrachlorophenol
IUPAC Name 2,3,4,5-tetrachlorophenol
SMILES Oc1cc(Cl)c(Cl)c(Cl)c1Cl
Specifications
Quantity 1 g
InChI Formula InChI=1S/C6H2Cl4O/c7-2-1-3(11)5(9)6(10)4(2)8/h1,11H
Formula Weight 229.886
Percent Purity >95%
Chemical Name or Material 2,3,4,5-Tetrachlorophenol
Grade HPLC