missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Thienyl)pyrrolidine Hydrochloride, TRC

Product Code. 30456357
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456357 500 mg 500mg
30454813 50 mg 50mg
30455122 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456357 Supplier Toronto Research Chemicals Supplier No. TRCT775935500MG
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2-Thienyl)pyrrolidine Hydrochloride
Molecular Formula C8 H11 N S . H Cl
Quantity 500 mg
InChI Formula InChI=1S/C8H11NS.ClH/c1-3-7(9-5-1)8-4-2-6-10-8;/h2,4,6-7,9H,1,3,5H2;1H
SMILES Cl.C1CNC(C1)c2cccs2
IUPAC Name 2-thiophen-2-ylpyrrolidine;hydrochloride
Molecular Weight (g/mol) 189.706
Formula Weight 189.038
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.