missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-[2-(tert-Butyldimethylsiloxy)ethyl]benzeneboronic acid, 96%

Product Code. 15450606
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450606 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15450606 Supplier Alfa Aesar™ Supplier No. H53339.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 913835-62-8
Melting Point 92°C to 94°C
Molecular Formula C14H25BO3Si
MDL Number MFCD08436049
Quantity 250 mg
Synonym 2-2-tert-butyldimethylsilyloxy ethyl phenylboronic acid, 2-2-tert-butyldimethylsilyloxy ethyl benzeneboronic acid, boronicacid, b-2-2-1,1-dimethylethyl dimethylsilyl oxy ethyl phenyl, 2-2-tert-butyldimethylsilyl oxy ethyl phenylboronic acid, 2-2-tert-butyldimethylsilyl oxy ethyl phenyl boronic acid, acmc-209r9p, 2-2-tert-butyldimethylsilyl-oxy ethyl phenyl boronic acid, 2-2-tert-butyldimethylsilyl oxy ethyl phenyl boronicacid, 2-2-tert-butyl dimethyl silyl oxy ethyl phenyl boronic acid, 2-2-1,1-dimethylethyl dimethylsilyl oxy ethyl phenyl boronic acid
InChI Key IYNGSWXPLWIRBQ-UHFFFAOYSA-N
SMILES B(C1=CC=CC=C1CCO[Si](C)(C)C(C)(C)C)(O)O
IUPAC Name [2-[2-[tert-butyl(dimethyl)silyl]oxyethyl]phenyl]boronic acid
Molecular Weight (g/mol) 280.246
PubChem CID 44119349
Formula Weight 280.25
Percent Purity 96%
Chemical Name or Material 2-[2-(tert-Butyldimethylsiloxy)ethyl]benzeneboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.