missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Naphthyl)-4-(5,6,7,8-tetrahydro-2-naphthyl)thiazole, 97%, Thermo Scientific™

Product Code. 15424216
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15424216 2 g 2g
15434216 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424216 Supplier Thermo Scientific Alfa Aesar Supplier No. H58405.04
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1421261-80-4
Molecular Formula C23H19NS
Molecular Weight (g/mol) 341.472
MDL Number MFCD22682973
InChI Key ISODJAPESPIWDI-UHFFFAOYSA-N
Synonym 2-naphthalen-2-yl-4-5,6,7,8-tetrahydronaphthalen-2-yl-1,3-thiazole, 2-2-naphthyl-4-5,6,7,8-tetrahydro-2-naphthyl thiazole
PubChem CID 97030349
IUPAC Name 2-naphthalen-2-yl-4-(5,6,7,8-tetrahydronaphthalen-2-yl)-1,3-thiazole
SMILES C1CCC2=C(C1)C=CC(=C2)C3=CSC(=N3)C4=CC5=CC=CC=C5C=C4

Specifications

Quantity 2 g
Formula Weight 341.47
Percent Purity 97%
Chemical Name or Material 2-(2-Naphthyl)-4-(5,6,7,8-tetrahydro-2-naphthyl)thiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.