Learn More
2,2'-Furil dioxime, mixture of isomers, 97%
Product Code. 11426856
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426856 5 g 5g
11416856 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11426856 Supplier Thermo Scientific Alfa Aesar Supplier No. B23972.06
 Restricted Item
In Stock
Add to basket
CAS: 522-27-0 | C10H8N2O4 | 220.18 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 522-27-0
Molecular Formula C10H8N2O4
Molecular Weight (g/mol) 220.18
MDL Number MFCD00075315
InChI Key RBOZTFPIXJBLPK-DSOJMZEYSA-N
Synonym alpha-furil dioxime, furildioxime, alpha-furyldioxime, di-2-furylglyoxime, bis 2-furyl glyoxime, di-2-furylethanedione dioxime, ethanedione, di-2-furanyl-, dioxime, glyoxime, di-2-furyl, furil, dioxime, 1z,2z-1,2-di furan-2-yl ethane-1,2-dione dioxime
PubChem CID 5359431
SMILES O\N=C(\C(=N/O)\C1=CC=CO1)/C1=CC=CO1
Specifications
Melting Point ∼165°C
Quantity 5 g
Beilstein 85894
Merck Index 14,4306
IUPAC Name N-[(E)-1,2-bis(furan-2-yl)-2-nitrosoethenyl]hydroxylamine
Formula Weight 220.19
Percent Purity 97%
Chemical Name or Material 2,2'-Furil dioxime, mixture of isomers