missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylglutaric acid, 98+%

Product Code. 11484467
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484467 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11484467 Supplier Thermo Scientific Alfa Aesar Supplier No. B20481.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 681-57-2 | C7H12O4 | 160.169 g/mol

2,2-Dimethylglutaric acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dimethylglutaric acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
It hydrolyzes with water.

Notes
Sto
TRUSTED_SUSTAINABILITY

Specifications

CAS 681-57-2
Melting Point 83°C to 85°C
Odor Odorless
Molecular Formula C7H12O4
MDL Number MFCD00004197
Quantity 25 g
Beilstein 1768501
Synonym 2,2-dimethylglutaric acid, 2,2-dimethylglutaricacid, unii-s46amq2g6c, pentanedioic acid, dimethyl, s46amq2g6c, glutaric acid, 2,2-dimethyl, .alpha.,.alpha.-dimethylglutaric acid, pentanedioic acid, 2,2-dimethyl, dimethylglutaric acid, dsstox_cid_1585
Solubility Information It hydrolyzes with water.
InChI Key BTUDGPVTCYNYLK-UHFFFAOYSA-N
SMILES CC(C)(CCC(=O)O)C(=O)O
IUPAC Name 2,2-dimethylpentanedioic acid
Molecular Weight (g/mol) 160.169
PubChem CID 12681
ChEBI CHEBI:68459
Formula Weight 160.17
Percent Purity ≥98%
Chemical Name or Material 2,2-Dimethylglutaric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.