missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Chloroethoxy)ethanol, Mikromol™

Product Code. 30049431
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049431 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049431 Supplier Mikromol Supplier No. MM0852.050100
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-(2-Chloroethoxy)ethanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2-Chloroethoxy)ethanol
CAS 628-89-7
Analyte or Component Names 2-(2-Chloroethoxy)ethanol
Shipping Condition Room Temperature
Molecular Formula C4 H9 Cl O2
Certifications/Compliance ISO 9001
Synonym Ethanol, 2-(2-chloroethoxy)-, 2-(2-Hydroxyethoxy)ethyl chloride, 2-(2'-Chloroethoxy)ethanol, 2-(Chloroethoxy)ethanol, 2-Chloroethyl 2-hydroxyethyl ether, 2-[(2-Chloroethyl)oxy]ethanol, 5-Chloro-3-oxa-1-pentanol, Diethylene glycol monochlorohydrin, Diglycol chlorohydrin, Ethylene glycol mono(2-chloroethyl) ether, NSC 2648
InChI Formula InChI=1S/C4H9ClO2/c5-1-3-7-4-2-6/h6H,1-4H2
SMILES OCCOCCCl
IUPAC Name 2-(2-chloroethoxy)ethanol
Molecular Weight (g/mol) 124.566
Quantity 100 mg
Formula Weight 124.0291 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.