Learn More
2,2'-Bis(hydroxyphenyl)methane, TRC
Specifications
Specifications
| Chemical Name or Material | 2,2'-Bisphenol F |
| CAS | 2467-02-9 |
| Molecular Formula | C13 H12 O2 |
| Quantity | 1 g |
| Synonym | Phenol, 2,2'-methylenedi- (7CI,8CI), 2,2'-Methylenebis[phenol], 2,2'-Bis(hydroxyphenyl)methane, 2,2'-Dihydroxydiphenylmethane, 2,2'-Methylenediphenol, Bis(2-hydroxyphenyl)methane, Bisphenol F, NSC 402103, o,o'-Bis(hydroxyphenyl)methane, o-(o-Hydroxybenzyl)phenol |
| InChI Formula | InChI=1S/C13H12O2/c14-12-7-3-1-5-10(12)9-11-6-2-4-8-13(11)15/h1-8,14-15H,9H2 |
| SMILES | Oc1ccccc1Cc2ccccc2O |
| IUPAC Name | 2-[(2-hydroxyphenyl)methyl]phenol |
| Molecular Weight (g/mol) | 200.23 |
| Formula Weight | 200.0837 |