missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, 98%

Product Code. p-7030054
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437705 250 mg 250mg
11446490 1 g 1g
11456490 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15437705 Supplier Thermo Scientific Alfa Aesar Supplier No. H27585.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 153305-67-0 | C48H40P2 | 678.796 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153305-67-0
Molecular Formula C48H40P2
Molecular Weight (g/mol) 678.796
MDL Number MFCD01311709
InChI Key IOPQYDKQISFMJI-UHFFFAOYSA-N
Synonym r-tol-binap, s-tol-binap, s---2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, 2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-t-binap, rac-tol-binap, r-+-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, tol-binap, s-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-p-tol-binap
PubChem CID 4189906
IUPAC Name [1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane
SMILES CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C

Specifications

Melting Point 267°C to 271°C
Quantity 250 mg
Formula Weight 678.79
Percent Purity 98%
Chemical Name or Material (+/-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.