missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(chloromethyl)-1,3-propanediol bis(bis(2-chloroethyl) phosphate), Dr. Ehrenstorfer

Product Code. 30325009
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325009 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325009 Supplier Dr. Ehrenstorfer Supplier No. DREC10651100
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Antiblaze V6
CAS 38051-10-4
Analyte or Component Names Antiblaze V6
Shipping Condition +20°C
Molecular Formula C13 H24 Cl6 O8 P2
Certifications/Compliance ISO/IEC 17025
Synonym Phosphoric acid, 2,2-bis(chloromethyl)-1,3-propanediyl tetrakis(2-chloroethyl) ester (9CI), Phosphoric acid, bis(2-chloroethyl) ester, diester with 2,2-bis(chloromethyl)-1,3-propanediol (7CI), 2,2-Bis(chloromethyl)-1,3-propanediol 2-chloroethanol phosphate (1:4:2), 2,2-Bis(chloromethyl)-1,3-propanediyl bis[bis(2-chloroethyl) phosphate], 2,2-Bis(chloromethyl)trimethylene bis[bis(2-chloroethyl) phosphate], 2,2-Bis-(chloromethyl)-propane-1,3-diyltetrakis(2-chloroethyl) bisphosphate, Amgard V 6, Antiblaze 100, Antiblaze AB 100, Antiblaze V 6, Celltech 60, Phosgard 2XC20, Tetrakis(2-chlorethyl)dichloroisopentyldiphosphate, Tolgard V 6, V 6, V 6 (fireproofing agent)
InChI Formula InChI=1S/C13H24Cl6O8P2/c14-1-5-22-28(20,23-6-2-15)26-11-13(9-18,10-19)12-27-29(21,24-7-3-16)25-8-4-17/h1-12H2
SMILES ClCCOP(=O)(OCCCl)OCC(CCl)(CCl)COP(=O)(OCCCl)OCCCl
IUPAC Name [2-[bis(2-chloroethoxy)phosphoryloxymethyl]-3-chloro-2-(chloromethyl)propyl] bis(2-chloroethyl) phosphate
Molecular Weight (g/mol) 582.99
Quantity 10 mg
Formula Weight 579.9078 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.