missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Azobis[2,4-dimethylvaleronitrile], TRC

Product Code. 30009481
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009481 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30009481 Supplier Toronto Research Chemicals Supplier No. TRCA932525100G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4419-11-8
Molecular Formula C14H24N4
Quantity 100 g
Synonym 2,2'-(1,2-Diazenediyl)bis[2,4-dimethylpentanenitrile], 2,2'-Azobis(dimethylvaleronitrile), 2,2'-Azobis[2,4-dimethylpentanenitrile], 2,2'-Azodi(2,4-dimethylvaleronitrile), ABVN, ADVN, AIVN, AMVN, V 65, V 65B, Vazo 52, alpha,alpha'-Azobis(dimethylvaleronitrile), alpha,alpha'-Azobis(alpha,gamma-dimethylvaleronitrile), alpha,alpha'-Azobis[2,4-dimethylvaleronitrile];
InChI Formula InChI=1S/C14H24N4/c1-11(2)7-13(5,9-15)17-18-14(6,10-16)8-12(3)4/h11-12H,7-8H2,1-6H3
SMILES CC(C)CC(C)(N=NC(C)(CC(C)C)C#N)C#N
IUPAC Name 2-[(E)-(2-cyano-4-methylpentan-2-yl)diazenyl]-2,4-dimethylpentanenitrile
Molecular Weight (g/mol) 248.37
Formula Weight 248.2
Percent Purity >95%
Chemical Name or Material 2,2'-Azobis[2,4-dimethylvaleronitrile]
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.