missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Aminoethyl)pyridine, MP Biomedicals

Product Code. 11417275 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11417275 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11417275 Supplier MP Biomedicals Supplier No. 0520936910
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 2706-56-1
Molecular Formula C7H10N2
MDL Number MFCD00006367
Quantity 10 g
Synonym 2-2-aminoethyl pyridine, 2-pyridylethylamine, 2-2-pyridyl ethylamine, 2-pyridin-2-yl ethanamine, 2-pyridineethanamine, demethylbetahistine, 2-pyridin-2-yl ethan-1-amine, alpha-pyridylethylamine, pyridine, 2-2-aminoethyl, 2-aminoethylpyridine
InChI Key XPQIPUZPSLAZDV-UHFFFAOYSA-N
SMILES NCCC1=CC=CC=N1
IUPAC Name 2-(pyridin-2-yl)ethan-1-amine
Molecular Weight (g/mol) 122.17
PubChem CID 75919
ChEBI CHEBI:74024
Formula Weight 122.17
Chemical Name or Material 2-(2-Aminoethyl)pyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.