Learn More
2,2,5-Trimethyl-1,3-dioxane-5-carboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2,2,5-Trimethyl-1,3-dioxane-5-carboxylic Acid |
| CAS | 16837-14-2 |
| Molecular Formula | C8 H14 O4 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C8H14O4/c1-7(2)11-4-8(3,5-12-7)6(9)10/h4-5H2,1-3H3,(H,9,10) |
| SMILES | CC1(C)OCC(C)(CO1)C(=O)O |
| IUPAC Name | 2,2,5-trimethyl-1,3-dioxane-5-carboxylic acid |
| Molecular Weight (g/mol) | 174.19 |
| Formula Weight | 174.0892 |