Learn More
2,2',4,4'-Tetrahydro-5,5'-dimethyl-2,2'-bis(4-methylphenyl)[4,4'-bi-3H-pyrazole]-3,3'-dione, TRC
Specifications
Specifications
| Chemical Name or Material | 2,2',4,4'-Tetrahydro-5,5'-dimethyl-2,2'-bis(4-methylphenyl)[4,4'-bi-3H-pyrazole]-3,3'-dione |
| CAS | 102827-44-1 |
| Molecular Formula | C22 H22 N4 O2 |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C22H22N4O2/c1-13-5-9-17(10-6-13)25-21(27)19(15(3)23-25)20-16(4)24-26(22(20)28)18-11-7-14(2)8-12-18/h5-12,19-20H,1-4H3 |
| SMILES | CC1=NN(C(=O)C1C2C(=NN(C2=O)c3ccc(C)cc3)C)c4ccc(C)cc4 |
| IUPAC Name | 5-methyl-4-[3-methyl-5-oxo-1-(p-tolyl)-4H-pyrazol-4-yl]-2-(p-tolyl)-4H-pyrazol-3-one |
| Molecular Weight (g/mol) | 375.44 |
| Formula Weight | 375.1821 |