Learn More
2,2',4,4'-Tetrahydro-5,5'-dimethyl-2,2'-bis(4-methylphenyl)[4,4'-bi-3H-pyrazole]-3,3'-dione, TRC
Product Code. 30449500
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449500 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30449500 Supplier Toronto Research Chemicals Supplier No. TRCT890520100MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,2',4,4'-Tetrahydro-5,5'-dimethyl-2,2'-bis(4-methylphenyl)[4,4'-bi-3H-pyrazole]-3,3'-dione
CAS 102827-44-1
Molecular Formula C22 H22 N4 O2
Quantity 100 mg
InChI Formula InChI=1S/C22H22N4O2/c1-13-5-9-17(10-6-13)25-21(27)19(15(3)23-25)20-16(4)24-26(22(20)28)18-11-7-14(2)8-12-18/h5-12,19-20H,1-4H3
SMILES CC1=NN(C(=O)C1C2C(=NN(C2=O)c3ccc(C)cc3)C)c4ccc(C)cc4
IUPAC Name 5-methyl-4-[3-methyl-5-oxo-1-(p-tolyl)-4H-pyrazol-4-yl]-2-(p-tolyl)-4H-pyrazol-3-one
Molecular Weight (g/mol) 375.44
Formula Weight 375.1821