Learn More
2,2,3,3-Tetramethoxybutane, 95%
Product Code. 15204268 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15204268 5 g 5g
15294258 1 g 1g
15214268 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15204268 Supplier Thermo Scientific Acros Supplier No. 450930050
Request Bulk or Custom Format
CAS: 176798-33-7 | C8H18O4 | 178.23 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 176798-33-7
Molecular Formula C8H18O4
Molecular Weight (g/mol) 178.23
MDL Number MFCD25458126
InChI Key COEBCOIFCLAOFZ-UHFFFAOYSA-N
Synonym butane, 2,2,3,3-tetramethoxy
PubChem CID 10856041
IUPAC Name 2,2,3,3-tetramethoxybutane
SMILES COC(C)(OC)C(C)(OC)OC
Specifications
Boiling Point 172.0°C to 174.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 178.23
Percent Purity 95%
Chemical Name or Material 2,2,3,3-Tetramethoxybutane
compliance-icons
Product Identifier
  • 2,2,3,3-Tetramethoxybutane, 95%
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.