missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethylamine hydrochloride, 98%

Product Code. p-7022229
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454130 10 g 10g
11464130 50 g 50g
11492707 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454130 Supplier Thermo Scientific Alfa Aesar Supplier No. A14904.09
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 373-88-6 | C2H5ClF3N | 135.51 g/mol

2,2,2-Trifluoroethylamine hydrochloride, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trifluoroethylamine hydrochloride, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff

Solubility
It is Soluble in 0.7 to water, but soluble in ethanol and chloroform, slightly soluble in benzene, hardly soluble in ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Keep away from strong oxidizing agents, acids, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 373-88-6
Molecular Formula C2H5ClF3N
Molecular Weight (g/mol) 135.51
MDL Number MFCD00012875
InChI Key ZTUJDPKOHPKRMO-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethylamine hydrochloride, 2,2,2-trifluoroethanamine hydrochloride, trifluoroethylamine hydrochloride, ethanamine, 2,2,2-trifluoro-, hydrochloride, 2,2,2-trifluoroethylaminehydrochloride, 2,2,2-trifluoroethylamine, hydrochloride, 2,2,2-trifluoroethan-1-amine hydrochloride, 2,2,2-trifluoroethylamine hcl, ethylamine, trifluoro-, hydrochloride, ethanamine, 2,2,2-trifluoro-, hydrochloride 1:1
PubChem CID 9772
IUPAC Name 2,2,2-trifluoroethanamine;hydrochloride
SMILES [H+].[Cl-].NCC(F)(F)F

Specifications

Melting Point 218°C (sublimation)
Linear Formula CF3CH2NH2·HCl
Quantity 10 g
Beilstein 3652103
Sensitivity Hygroscopic
Solubility Information It is Soluble in 0.7 to water,but soluble in ethanol and chloroform,slightly soluble in benzene,hardly soluble in ether.
Formula Weight 135.52
Percent Purity 98%
Chemical Name or Material 2,2,2-Trifluoroethylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.