Learn More
2,2,2-Trifluoro-N-[2-(4-iodo-2,5-dimethoxyphenyl)ethyl]acetamide, TRC
Product Code. 30456692
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456692 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30456692 Supplier Toronto Research Chemicals Supplier No. TRCT783065250MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,2,2-Trifluoro-N-[2-(4-iodo-2,5-dimethoxyphenyl)ethyl]acetamide
CAS 159277-10-8
Molecular Formula C12H13F3INO3
Quantity 250 mg
InChI Formula InChI=1S/C12H13F3INO3/c1-19-9-6-8(16)10(20-2)5-7(9)3-4-17-11(18)12(13,14)15/h5-6H,3-4H2,1-2H3,(H,17,18)
SMILES COc1cc(CCNC(=O)C(F)(F)F)c(OC)cc1I
IUPAC Name 2,2,2-trifluoro-N-[2-(4-iodo-2,5-dimethoxyphenyl)ethyl]acetamide
Molecular Weight (g/mol) 403.14
Formula Weight 402.9892