missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 15458175
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15458175 250 mg 250mg
15448175 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458175 Supplier Thermo Scientific Alfa Aesar Supplier No. H32717.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1138011-22-9
Molecular Formula C7H8ClF3N2
Molecular Weight (g/mol) 212.6
MDL Number MFCD06739079
InChI Key NLDVAGWIUPSGQC-UHFFFAOYSA-N
Synonym 2,2,2-trifluoro-1-pyridin-3-yl ethanamine hydrochloride, +/--2,2,2-trifluoro-1-pyridin-3-ylethylamine hydrochloride, 2,2,2-trifluoro-1-pyridin-3-ylethylamine hydrochloride, 2,2,2-trifluoro-1-pyridin-3-yl-ethylamine hydrochloride, 2,2,2-trifluoro-1-3-pyridyl ethylamine hydrochloride, acmc-20aokz, 2,2,2-trifluoro-1-3-pyridinyl ethanamine hydrochloride, 2,2,2-trifluoro-1-3-pyridinyl ethylamine hydrochloride, 2,2,2-tris fluoranyl-1-pyridin-3-yl-ethanamine hydrochloride
PubChem CID 45594310
IUPAC Name 2,2,2-trifluoro-1-pyridin-3-ylethanamine;hydrochloride
SMILES C1=CC(=CN=C1)C(C(F)(F)F)N.Cl

Specifications

Melting Point 97°C to 102°C
Quantity 250 mg
Formula Weight 212.6
Percent Purity 95%
Chemical Name or Material 2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.