missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Crypt, TRC

Product Code. 30007475
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007475 10 mg 10mg
30007763 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007475 Supplier Toronto Research Chemicals Supplier No. TRCC82746010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23978-09-8
Molecular Formula C18 H36 N2 O6
Molecular Weight (g/mol) 376.49
Synonym 4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane, 2,2,2-Crypt, 2,2,2-Cryptand, 2,2,2-Cryptate, Crypt-2,2,2, Cryptand 222, Cryptand C 222, Cryptand[2.2.2], Cryptate 222, Cryptating agent 222, Cryptofix 222, K 222, Kryptand 222, Kryptofix 222, NSC 264495, WS-A 101, [2,2,2]Crypand
IUPAC Name 4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane
SMILES C1COCCN2CCOCCOCCN(CCOCCOCC2)CCO1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H36N2O6/c1-7-21-13-14-24-10-4-20-5-11-25-17-15-22-8-2-19(1)3-9-23-16-18-26-12-6-20/h1-18H2
Formula Weight 376.2573
Chemical Name or Material 4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.