Learn More
2-[2-(2-Amino-9H-purin-9-yl)ethyl]propane-1,3-diol Hydrochloride, Mikromol™
Product Code. 30052150
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052150 100 mg 100mg
30050183 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30052150 Supplier Mikromol Supplier No. MM0620.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-[2-(2-Amino-9H-purin-9-yl)ethyl]propane-1,3-diol Hydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-[2-(2-Amino-9H-purin-9-yl)ethyl]propane-1,3-diol hydrochloride
CAS 246021-75-0
Analyte or Component Names 2-[2-(2-Amino-9H-purin-9-yl)ethyl]propane-1,3-diol hydrochloride
Shipping Condition Room Temperature
Molecular Formula C10 H15 N5 O2 . Cl H
Certifications/Compliance ISO 9001
Synonym 1,3-Propanediol, 2-[2-(2-amino-9H-purin-9-yl)ethyl]-, hydrochloride (1:1), 1,3-Propanediol, 2-[2-(2-amino-9H-purin-9-yl)ethyl]-, monohydrochloride (9CI), Famciclovir Related Compound A as Hydrochloride
InChI Formula InChI=1S/C10H15N5O2.ClH/c11-10-12-3-8-9(14-10)15(6-13-8)2-1-7(4-16)5-17;/h3,6-7,16-17H,1-2,4-5H2,(H2,11,12,14);1H
SMILES Cl.Nc1ncc2ncn(CCC(CO)CO)c2n1
IUPAC Name 2-[2-(2-aminopurin-9-yl)ethyl]propane-1,3-diol;hydrochloride
Molecular Weight (g/mol) 273.72
Quantity 100 mg
Formula Weight 273.0993 g/mol