Learn More
2-[2-(2-Amino-9H-purin-9-yl)ethyl]propane-1,3-diol Hydrochloride, Mikromol™
Description
2-[2-(2-Amino-9H-purin-9-yl)ethyl]propane-1,3-diol Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 2-[2-(2-Amino-9H-purin-9-yl)ethyl]propane-1,3-diol hydrochloride |
| CAS | 246021-75-0 |
| Analyte or Component Names | 2-[2-(2-Amino-9H-purin-9-yl)ethyl]propane-1,3-diol hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H15 N5 O2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1,3-Propanediol, 2-[2-(2-amino-9H-purin-9-yl)ethyl]-, hydrochloride (1:1), 1,3-Propanediol, 2-[2-(2-amino-9H-purin-9-yl)ethyl]-, monohydrochloride (9CI), Famciclovir Related Compound A as Hydrochloride |
| InChI Formula | InChI=1S/C10H15N5O2.ClH/c11-10-12-3-8-9(14-10)15(6-13-8)2-1-7(4-16)5-17;/h3,6-7,16-17H,1-2,4-5H2,(H2,11,12,14);1H |
| SMILES | Cl.Nc1ncc2ncn(CCC(CO)CO)c2n1 |
| IUPAC Name | 2-[2-(2-aminopurin-9-yl)ethyl]propane-1,3-diol;hydrochloride |
| Show More |