Learn More
2-[[2-(2,2,2-Trifluoroethoxy)phenoxy]methyl]-1,3-dioxolane, TRC
Product Code. 30455630
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455630 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455630 Supplier Toronto Research Chemicals Supplier No. TRCT79018550MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-((2-(2,2,2-Trifluoroethoxy)phenoxy)methyl)-1,3-dioxolane
CAS 1269801-74-2
Molecular Formula C27 H24 O8
Quantity 50 mg
InChI Formula InChI=1S/C27H24O8/c1-31-27-23(35-26(30)20-15-9-4-10-16-20)22(34-25(29)19-13-7-3-8-14-19)21(33-27)17-32-24(28)18-11-5-2-6-12-18/h2-16,21-23,27H,17H2,1H3/t21-,22?,23?,27-/m1/s1
SMILES CO[C@@H]1O[C@H](COC(=O)c2ccccc2)C(OC(=O)c3ccccc3)C1OC(=O)c4ccccc4
Molecular Weight (g/mol) 476.475
Formula Weight 476.147