missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(1,3,3-Trimethyl-2-indolinylidene)ethylidene]-1-tetralone, predominantly trans/trans, 96%, Alfa Aesar™

Product Code. 15435205
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435205 250 mg 250mg
15425205 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435205 Supplier Thermo Scientific Alfa Aesar Supplier No. H34300.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 53704-25-9
Molecular Formula C23H23NO
Molecular Weight (g/mol) 329.443
MDL Number MFCD06409585
InChI Key DTPLBBMEWNNVFO-UHFFFAOYSA-N
Synonym e-2-2-e-1,3,3-trimethylindolin-2-ylidene ethylidene-3,4-dihydronaphthalen-1 2h-one, 2e-2-2e-2-1,3,3-trimethyl-1,3-dihydro-2h-indol-2-ylidene ethylidene-3,4-dihydronaphthalen-1 2h-one, 2-2-1,3,3-trimethylindol-2-ylidene ethylidene-3,4-dihydronaphthalen-1-one, 2-2-1,3,3-trimethyl-1,3-dihydro-2h-indol-2-ylidene ethylidene-3,4-dihydronaphthalen-1 2h-one
PubChem CID 53426997
IUPAC Name 2-[2-(1,3,3-trimethylindol-2-ylidene)ethylidene]-3,4-dihydronaphthalen-1-one
SMILES CC1(C2=CC=CC=C2N(C1=CC=C3CCC4=CC=CC=C4C3=O)C)C

Specifications

Melting Point 210°C to 214°C
Quantity 250 mg
Formula Weight 329.25
Percent Purity 96%
Chemical Name or Material 2-[2-(1,3,3-Trimethyl-2-indolinylidene)ethylidene]-1-tetralone, predominantly trans/trans
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.