Learn More
2-(1-Methylpiperidin-3-yl)ethan-1-ol, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(1-Methylpiperidin-3-yl)ethan-1-ol |
| CAS | 101257-32-3 |
| Molecular Formula | C8H17NO |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C8H17NO/c1-9-5-2-3-8(7-9)4-6-10/h8,10H,2-7H2,1H3 |
| SMILES | CN1CCCC(CCO)C1 |
| IUPAC Name | 2-(1-methylpiperidin-3-yl)ethanol |
| Molecular Weight (g/mol) | 143.23 |
| Formula Weight | 143.131 |