Learn More
2-(1-Methylethyl)-6-propylphenol, Mikromol™
Product Code. 30049691
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049691 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30049691 Supplier Mikromol Supplier No. MM0461.01
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-(1-Methylethyl)-6-propylphenol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Isopropyl-6-propylphenol
CAS 74663-48-2
Analyte or Component Names 2-Isopropyl-6-propylphenol
Shipping Condition Room Temperature
Molecular Formula C12 H18 O
Certifications/Compliance ISO 9001
Synonym 2-(1-Methylethyl)-6-propylphenol, Propofol Imp. O (EP), Propofol Impurity O
InChI Formula InChI=1S/C12H18O/c1-4-6-10-7-5-8-11(9(2)3)12(10)13/h5,7-9,13H,4,6H2,1-3H3
SMILES CCCc1cccc(C(C)C)c1O
IUPAC Name 2-propan-2-yl-6-propylphenol
Molecular Weight (g/mol) 178.27
Quantity 50 mg
Formula Weight 178.1358 g/mol