missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,2S)-(+)-2-Benzyloxycyclopentylamine, ChiPros 99+%, ee 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
(1S,2S)-(+)-2-Benzyloxycyclopentylamine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Chiral amines play an important role in stereoselective organic synthesis. They are used directly as resolving agents, building blocks or chiral auxiliaries.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under inert gas. It is sensitive to air and carbon dioxide.
(1S,2S)-(+)-2-Benzyloxycyclopentylamine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Chiral amines play an important role in stereoselective organic synthesis. They are used directly as resolving agents, building blocks or chiral auxiliaries.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under inert gas. It is sensitive to air and carbon dioxide.
Specifications
Specifications
| Melting Point | 23°C |
| Boiling Point | 90°C to 93°C (0.23 mmHg) |
| Flash Point | >110°C (230°F) |
| Quantity | 1 g |
| UN Number | UN2735 |
| Synonym | 1s,2s-1-amino-2-benzyloxycyclopentane, 1s,2s-2-benzyloxy cyclopentanamine, 1s,2s-2-benzyloxycyclopentylamine, 1s,2s-+-2-benzyloxycyclopentylamine, 1s,2s-2-benzyloxy cyclopentan-1-amine, 1s,25-2-benzyloxycyclopentylamine, 1s,2s-2-benzyloxycyclopentanamine, 1s, 2s-2-benzyloxycyclopentylamine, 1s,2s-2-benzyloxy-cyclopentylamine, 1s,2s-2-benzyloxycyclopentyl-amine |
| Optical Rotation | +57.5° (Neat) |
| Formula Weight | 191.27 |
| Percent Purity | ≥99% |
| Assay | ee 99% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction