CAS RN 181657-57-8
CAS RN 181657-57-8
IUPAC Name:
2-(benzyloxy)cyclopentan-1-amine
Synonyms:
2-(benzyloxy)cyclopentan-1-amine
Molecular Weight (g/mol):
191.27
Molecular Formula:
C12H17NO
InChi Key:
JIMSXLUBRRQALI-UHFFFAOYNA-N
SMILES:
NC1CCCC1OCC1=CC=CC=C1
1
–
1
of
1
results
1
(1S,2S)-(+)-2-Benzyloxycyclopentylamine, ChiPros 99+%, ee 99%, Thermo Scientific™
CAS: 181657-57-8 Molecular Formula: C12H17NO Molecular Weight (g/mol): 191.274 MDL Number: MFCD01075751 InChI Key: JIMSXLUBRRQALI-RYUDHWBXSA-N PubChem CID: 11908836 IUPAC Name: (1S,2S)-2-phenylmethoxycyclopentan-1-amine SMILES: C1CC(C(C1)OCC2=CC=CC=C2)N