missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-2-Aminocyclohexanecarboxylic acid, 97%

Product Code. 15479517
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15479517 5 g 5g
15490116 250 mg 250mg
15469517 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15479517 Supplier Thermo Scientific Alfa Aesar Supplier No. H52779.06
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 24716-93-6 | C7H13NO2 | 143.186 g/mol

(1S,2S)-2-Aminocyclohexanecarboxylic acid is used in the preparation of enantiomerically pure beta-amino acids. It is also involved in the synthesis of mono and dihydroxylated derivatives of 2-aminocyclohexanecarboxylic acid by Diels-Alder adduct of ethyl(E)-3-nitroacrylate and furan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S,2S)-2-Aminocyclohexanecarboxylic acid is used in the preparation of enantiomerically pure beta-amino acids. It is also involved in the synthesis of mono and dihydroxylated derivatives of 2-aminocyclohexanecarboxylic acid by Diels-Alder adduct of ethyl(E)-3-nitroacrylate and furan.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24716-93-6
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD01318270
InChI Key USQHEVWOPJDAAX-WDSKDSINSA-N
PubChem CID 7128323
IUPAC Name (1S,2S)-2-aminocyclohexane-1-carboxylic acid
SMILES C1CCC(C(C1)C(=O)O)N

Specifications

Synonym 1s,2s-2-aminocyclohexanecarboxylic acid, 1s,2s-2-aminocyclohexane-1-carboxylic acid, cyclohexanecarboxylic acid, 2-amino-, 1s,2s, 1s,2s-2-aminocyclohexanecarboxylicacid, 1s,2s-2-amino-cyclohexanecarboxylic acid, 2beta-amino-1alpha-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid,2-amino-, 1s,2s, 24716-92-5 1s,2s-2-aminocyclohexane-1-carboxylic acid
Solubility Information Soluble in water.
Quantity 5 g
Formula Weight 143.19
Optical Rotation +77° (c=1 in Water)
Percent Purity 97%
Chemical Name or Material (1S,2S)-2-Aminocyclohexanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.