Learn More
(1RS)-1-[1-(4-Chlorophenyl)cyclobutyl]-N,3-dimethylbutan-1-amine Hydrochloride (N-Monodesmethylsibutramine Hydrochloride), Mikromol™
Description
(1RS)-1-[1-(4-Chlorophenyl)cyclobutyl]-N,3-dimethylbutan-1-amine Hydrochloride (N-Monodesmethylsibutramine Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | Desmethylsibutramine Hydrochloride |
| CAS | 84467-94-7 |
| Analyte or Component Names | Desmethylsibutramine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H24 Cl N . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | (1RS)-1-[1-(4-Chlorophenyl)cyclobutyl]-N,3-dimethylbutan-1-amine Hydrochloride, Sibutramine Hydrochloride Monohydrate Imp. D (Pharmeuropa) as Hydrochloride, N-Monodesmethylsibutramine Hydrochloride, Desmethylsibutramine Hydrochloride, Sibutramine Related Compound D as Hydrochloride, Sibutramine Hydrochloride Monohydrate Impurity D, Sibutramine Impurity D |
| InChI Formula | InChI=1S/C16H24ClN.ClH/c1-12(2)11-15(18-3)16(9-4-10-16)13-5-7-14(17)8-6-13;/h5-8,12,15,18H,4,9-11H2,1-3H3;1H |
| SMILES | Cl.CNC(CC(C)C)C1(CCC1)c2ccc(Cl)cc2 |
| IUPAC Name | 1-[1-(4-chlorophenyl)cyclobutyl]-N,3-dimethylbutan-1-amine;hydrochloride |
| Show More |