missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(-)-1,2-Diaminocyclohexane, 98%

Product Code. p-7046357
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398935 250 mg 250mg
11378935 1 g 1g
11388935 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11398935 Supplier Thermo Scientific Alfa Aesar Supplier No. L14059.MD
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20439-47-8 | C6H14N2 | 114.19 g/mol

(1R,2R)-(-)-1,2-Diaminocyclohexane is a versatile ligand for the formation of metal complexes. Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R,2R)-(-)-1,2-Diaminocyclohexane is a versatile ligand for the formation of metal complexes. Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis.

Solubility
Soluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, carbondioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20439-47-8
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00062985,MFCD00063747
InChI Key SSJXIUAHEKJCMH-UHFFFAOYNA-N
Synonym 1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine
PubChem CID 43806
SMILES NC1CCCCC1N

Specifications

Melting Point 40°C to 43°C
Boiling Point 104°C to 110°C (40 mmHg)
Flash Point 70°C (158°F)
Odor Amine-like
Quantity 250 mg
UN Number UN3259
Beilstein 4780911
Optical Rotation −25.5° (c=5 in 1N HCl)
IUPAC Name cyclohexane-1,2-diamine
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material (1R,2R)-(-)-1,2-Diaminocyclohexane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.