Learn More
(1H-Imidazol-1-yl)(phenyl)methanone, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 10364-94-0 |
| Molecular Formula | C10H8N2O |
| MDL Number | MFCD00014499 |
| Quantity | 1 g |
| SMILES | O=C(N1C=NC=C1)C2=CC=CC=C2 |
| Molecular Weight (g/mol) | 172.18 |
| Percent Purity | 95% |
| Chemical Name or Material | (1H-Imidazol-1-yl)(phenyl)methanone |