Learn More
1H,1H,2H,2H-Perfluorooctyl-1-thiol, TRC
Specifications
Specifications
| Chemical Name or Material | 1H,1H,2H,2H-Perfluorooctyl-1-thiol |
| CAS | 34451-26-8 |
| Purity Grade Notes | GC |
| Molecular Formula | C8H5F13S |
| Quantity | 500 mg |
| Synonym | 1-Octanethiol','3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro- (9CI','ACI): 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanethiol (ACI), 1,1,2,2-Tetrahydroperfluorooctanethiol, 1,1,2,2-Tetrahydroperfluorooctyl mercaptan, 1H,1H,1H,2H-Perfluorooctanethiol, 1H,1H,2H,2H-Perfluorooctanethiol, 2-(Perfluorohexyl)ethane-1-thiol, 2-(Perfluorohexyl)ethanethiol, Foralkyl EM 6 |
| InChI Formula | InChI=1S/C8H5F13S/c9-3(10,1-2-22)4(11,12)5(13,14)6(15,16)7(17,18)8(19,20)21/h22H,1-2H2 |
| SMILES | FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCS |
| IUPAC Name | 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctane-1-thiol |
| Molecular Weight (g/mol) | 380.17 |
| Show More |