Learn More
19-Norandrosterone (d5 major), TRC
Specifications
Specifications
| Chemical Name or Material | 19-Norandrosterone-d5 |
| CAS | 1011714-16-1 |
| Molecular Formula | C18H23D5O2 |
| Quantity | 1 mg |
| Synonym | 3α-Hydroxy- 5α-estran-17-one-d5, 19-Norandrosterone-d5, 3α-Hydroxy-5α-estran-17-one-d5, 5α-Estran-3α-ol-17-one-d5 |
| InChI Formula | InChI=1S/C18H28O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h11-16,19H,2-10H2,1H3/t11-,12+,13-,14+,15+,16-,18-/m0/s1/i3D2,10D2,12D |
| SMILES | [H][C@]12[C@@]([C@@](CCC3=O)([H])[C@]3(C)CC2)([H])CC[C@@]4([H])C([2H])([2H])[C@]([2H])(O)C([2H])([2H])C[C@@]41[H] |
| Molecular Weight (g/mol) | 281.44 |
| Formula Weight | 281.2403 |